siRNA Details
| Gene Symbol: Gm15984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040684.1 | - | 7 | 1108 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB213859.2 | - | 2 | 102 | Chr. 5 - 74726874 - 74728605 [+] on Build GRCm38 | |

Gm15984
Genomic Map