siRNA Details
| Gene Symbol: 1700006H21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045900.2 | 3 | 371 | |||
| GenBank mRNA | AK005677.1 | - | 3 | 372 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY705860.1 | - | 3 | 372 | Chr. 13 - 107687397 - 107692169 [+] on Build GRCm38 | |

1700006H21Rik
Genomic Map