siRNA Details
Gene Symbol: Gm6300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033591.2 | 3 | 1253 | |||
GenBank mRNA | AK143764.1 | - | 3 | 1256 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm6300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033591.2 | 3 | 1253 | |||
GenBank mRNA | AK143764.1 | - | 3 | 1256 |