siRNA Details
| Gene Symbol: Trpm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031944.1 | - | 649 | |||
| AK034340.1 | - | 877 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI736607.1 | - | 1 | 338 | Chr. 19 - 22992580 - 22993218 [+] on Build GRCm38 | |

Trpm3
Genomic Map