siRNA Details
| Gene Symbol: Gm4668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041339.1 | - | 1 | 87 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB590615.2 | - | 1 | 87 | Chr. 9 - 116189180 - 116192999 [+] on Build GRCm38 | |

Gm4668
Genomic Map