siRNA Details
Gene Symbol: Gm4668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041339.1 | - | 1, 2 | 220 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB590615.2 | - | 1, 2 | 220 | Chr. 9 - 116189180 - 116192999 [+] on Build GRCm38 |
