siRNA Details
| Gene Symbol: Gm4668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041339.1 | - | 4 | 863 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB227220.2 | - | 3 | 620 | Chr. 9 - 116190822 - 116194870 [+] on Build GRCm38 | |

Gm4668
Genomic Map