siRNA Details
| Gene Symbol: Gm4668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041339.1 | - | 4 | 924 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4668| Gene Symbol: Gm4668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041339.1 | - | 4 | 924 | ||