siRNA Details
Gene Symbol: Gm15987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045009.2 | 2 | 813 | |||
GenBank mRNA | AK040061.1 | - | 2 | 692 | ||
AK157579.1 | - | 813 | ||||
AK172082.1 | - | 2 | 795 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm15987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045009.2 | 2 | 813 | |||
GenBank mRNA | AK040061.1 | - | 2 | 692 | ||
AK157579.1 | - | 813 | ||||
AK172082.1 | - | 2 | 795 |