siRNA Details
| Gene Symbol: Gm15987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045009.2 | 2 | 880 | |||
| GenBank mRNA | AK040061.1 | - | 2 | 758 | ||
| AK157579.1 | - | 879 | ||||
| AK172082.1 | - | 2 | 862 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15987| Gene Symbol: Gm15987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045009.2 | 2 | 880 | |||
| GenBank mRNA | AK040061.1 | - | 2 | 758 | ||
| AK157579.1 | - | 879 | ||||
| AK172082.1 | - | 2 | 862 | |||