siRNA Details
| Gene Symbol: Gm15987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045009.2 | 2 | 700 | |||
| GenBank mRNA | AK040061.1 | - | 2 | 579 | ||
| AK157579.1 | - | 700 | ||||
| AK172082.1 | - | 2 | 682 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB635100.1 | - | 2 | 579 | Chr. 6 - 128952684 - 128974907 [-] on Build GRCm38 | |

Gm15987
Genomic Map