siRNA Details
| Gene Symbol: Gm12708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089375.1 | - | 3 | 633 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY749636.1 | - | 3 | 633 | Chr. 4 - 95193313 - 95209365 [+] on Build GRCm38 | |

Gm12708
Genomic Map