siRNA Details
| Gene Symbol: Gm16096 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089373.1 | - | 637 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY764361.1 | - | 3 | 531 | Chr. 9 - 40780740 - 40783425 [-] on Build GRCm38 | |

Gm16096
Genomic Map