siRNA Details
| Gene Symbol: Atf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137400.1 | - | 1 | 3141 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB130570.2 | - | 1 | 546 | Chr. 15 - 102590321 - 102591009 [-] on Build GRCm38 | |

Atf7
Genomic Map