siRNA Details
| Gene Symbol: 4930434J06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046274.1 | 7 | 1513 | |||
| GenBank mRNA | AK015316.1 | - | 7 | 1513 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA464707.1 | - | 586 | Chr. - - [-] on Build | ||

4930434J06Rik
Genomic Map