siRNA Details
| Gene Symbol: 4930434J06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046274.1 | 5, 6 | 1187 | |||
| GenBank mRNA | AK015316.1 | - | 5, 6 | 1187 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930434J06Rik| Gene Symbol: 4930434J06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046274.1 | 5, 6 | 1187 | |||
| GenBank mRNA | AK015316.1 | - | 5, 6 | 1187 | ||