siRNA Details
| Gene Symbol: 4930434J06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046274.1 | 2 | 227 | |||
| GenBank mRNA | AK015316.1 | - | 2 | 227 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714869.1 | - | 2 | 227 | Chr. 14 - 71856709 - 71859905 [+] on Build GRCm38 | |

4930434J06Rik
Genomic Map