siRNA Details
| Gene Symbol: Ascc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011671.1 | - | 1 | 232 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ944505.1 | - | 1 | 170 | Chr. 11 - 4683962 - 4684827 [+] on Build GRCm38 | |

Ascc2
Genomic Map