siRNA Details
| Gene Symbol: Ascc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011671.1 | - | 1 | 1692 | ||
| AK144613.1 | - | 862 | ||||
| BC024840.1 | - | 14 | 1795 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ascc2| Gene Symbol: Ascc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011671.1 | - | 1 | 1692 | ||
| AK144613.1 | - | 862 | ||||
| BC024840.1 | - | 14 | 1795 | |||