siRNA Details
| Gene Symbol: Atf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003630.1 | - | 951 | |||
| AK170250.1 | - | 1041 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atf7| Gene Symbol: Atf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003630.1 | - | 951 | |||
| AK170250.1 | - | 1041 | ||||