siRNA Details
| Gene Symbol: Atf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003630.1 | - | 1324 | |||
| AK170250.1 | - | 1414 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ109070.1 | - | 1 | 74 | Chr. 15 - 102528641 - 102529081 [-] on Build GRCm38 | |

Atf7
Genomic Map