siRNA Details
| Gene Symbol: BC055402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037990.1 | 6, 7 | 1110 | |||
| GenBank mRNA | BC055402.1 | - | 6, 7 | 1107 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC055402| Gene Symbol: BC055402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037990.1 | 6, 7 | 1110 | |||
| GenBank mRNA | BC055402.1 | - | 6, 7 | 1107 | ||