siRNA Details
| Gene Symbol: 2700033N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012325.1 | - | 1 | 24 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY711481.1 | - | 1 | 31 | Chr. 2 - 116066218 - 116067367 [-] on Build GRCm38 | |

2700033N17Rik
Genomic Map