siRNA Details
| Gene Symbol: Gm32857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC060994.1 | - | 4 | 1163 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm32857| Gene Symbol: Gm32857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC060994.1 | - | 4 | 1163 | ||