siRNA Details
| Gene Symbol: Gm32857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC060994.1 | - | 3 | 546 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB598493.1 | - | 3 | 549 | Chr. 14 - 44226920 - 44315513 [-] on Build GRCm38 | |

Gm32857
Genomic Map