siRNA Details
| Gene Symbol: Gm12348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158747.1 | - | 2 | 79 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY247364.1 | - | 2 | 71 | Chr. 11 - 100398121 - 100402252 [+] on Build GRCm38 | |

Gm12348
Genomic Map