siRNA Details
| Gene Symbol: 1700084F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016373.1 | - | 2 | 305 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716020.1 | - | 2 | 305 | Chr. 13 - 70022116 - 70028319 [-] on Build GRCm38 | |

1700084F23Rik
Genomic Map