siRNA Details
| Gene Symbol: 4930590A17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016377.1 | - | 6 | 805 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DV658617.1 | - | 6 | 732 | Chr. 16 - 87909341 - 87952054 [+] on Build GRCm38 | |

4930590A17Rik
Genomic Map