siRNA Details
Gene Symbol: 5830416I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045384.1 | 3 | 1935 | |||
GenBank mRNA | AK080054.1 | - | 1 | 327 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 5830416I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045384.1 | 3 | 1935 | |||
GenBank mRNA | AK080054.1 | - | 1 | 327 |