siRNA Details
| Gene Symbol: 5830449F15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018000.1 | - | 1 | 97 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717925.1 | - | 1 | 97 | Chr. 16 - 23280161 - 23280685 [+] on Build GRCm38 | |

5830449F15Rik
Genomic Map