siRNA Details
| Gene Symbol: Asb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008728.1 | - | 2 | 280 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708630.1 | - | 2 | 277 | Chr. 13 - 3634089 - 3634782 [+] on Build GRCm38 | |

Asb13
Genomic Map