siRNA Details
| Gene Symbol: Atxn7l1os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016151.1 | - | 688 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715754.1 | - | 4 | 688 | Chr. 12 - 33359294 - 33394760 [-] on Build GRCm38 | |

Atxn7l1os2
Genomic Map