siRNA Details
Gene Symbol: Atxn7l1os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016151.1 | - | 400 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715754.1 | - | 4 | 400 | Chr. 12 - 33359294 - 33394760 [-] on Build GRCm38 |
