siRNA Details
| Gene Symbol: Gm14936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136133.1 | - | 3 | 355 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720073.1 | - | 3 | 353 | Chr. X - 112982803 - 113009753 [-] on Build GRCm38 | |

Gm14936
Genomic Map