siRNA Details
| Gene Symbol: 5033430J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017208.1 | - | 1 | 1194 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5033430J17Rik| Gene Symbol: 5033430J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017208.1 | - | 1 | 1194 | ||