siRNA Details
| Gene Symbol: 9430068D22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020479.1 | - | 1 | 198 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721819.1 | - | 1 | 198 | Chr. 15 - 26644653 - 26645289 [-] on Build GRCm38 | |

9430068D22Rik
Genomic Map