siRNA Details
| Gene Symbol: Gm41183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK075677.1 | - | 2 | 345 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB609746.1 | - | 2 | 345 | Chr. 14 - 66338393 - 66341229 [-] on Build GRCm38 | |

Gm41183
Genomic Map