siRNA Details
| Gene Symbol: 9430065F17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020476.1 | - | 3 | 241 | ||
| AK021377.1 | - | 3 | 271 | |||
| BY126544.1 | - | 3 | 203 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9430065F17Rik| Gene Symbol: 9430065F17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020476.1 | - | 3 | 241 | ||
| AK021377.1 | - | 3 | 271 | |||
| BY126544.1 | - | 3 | 203 | |||