siRNA Details
| Gene Symbol: 1700022H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045488.1 | 1 | 23 | |||
| GenBank mRNA | AK006240.1 | - | 1 | 24 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706323.1 | - | 1 | 24 | Chr. 12 - 9565236 - 9570585 [-] on Build GRCm38 | |

1700022H16Rik
Genomic Map