siRNA Details
| Gene Symbol: 1700047K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006714.1 | - | 2 | 867 | ||
| AV247949.1 | - | 1 | 283 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700047K16Rik| Gene Symbol: 1700047K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006714.1 | - | 2 | 867 | ||
| AV247949.1 | - | 1 | 283 | |||