siRNA Details
| Gene Symbol: Gm4035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079953.1 | - | 3 | 349 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB214962.2 | - | 3 | 389 | Chr. 13 - 37421946 - 37435151 [-] on Build GRCm38 | |

Gm4035
Genomic Map