siRNA Details
| Gene Symbol: Zfp618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084535.1 | - | 1 | 2172 | ||
| AK085069.1 | - | 1 | 3257 | |||
| BC059079.1 | - | 1 | 3717 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp618| Gene Symbol: Zfp618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084535.1 | - | 1 | 2172 | ||
| AK085069.1 | - | 1 | 3257 | |||
| BC059079.1 | - | 1 | 3717 | |||