siRNA Details
Gene Symbol: Zfp618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK084535.1 | - | 1 | 62 | ||
AK085069.1 | - | 1 | 1142 | |||
BC059079.1 | - | 1 | 1606 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK084535.1 | - | 1 | 62 | ||
AK085069.1 | - | 1 | 1142 | |||
BC059079.1 | - | 1 | 1606 |