siRNA Details
| Gene Symbol: Gm19782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045071.1 | 3 | 277 | |||
| GenBank mRNA | AK139312.1 | - | 3 | 279 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB591178.2 | - | 3 | 279 | Chr. 15 - 69924233 - 70074094 [+] on Build GRCm38 | |

Gm19782
Genomic Map