siRNA Details
| Gene Symbol: C030013C21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021072.1 | - | 1 | 272 | ||
| AV253635.2 | - | 1 | 353 | |||
| BB614470.1 | - | 1 | 492 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C030013C21Rik| Gene Symbol: C030013C21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021072.1 | - | 1 | 272 | ||
| AV253635.2 | - | 1 | 353 | |||
| BB614470.1 | - | 1 | 492 | |||