siRNA Details
Gene Symbol: C030013C21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021072.1 | - | 1 | 148 | ||
AV253635.2 | - | 1 | 229 | |||
BB614470.1 | - | 1 | 368 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C030013C21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021072.1 | - | 1 | 148 | ||
AV253635.2 | - | 1 | 229 | |||
BB614470.1 | - | 1 | 368 |