siRNA Details
| Gene Symbol: C030013C21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021072.1 | - | 1 | 205 | ||
| AV253635.2 | - | 1 | 286 | |||
| BB614470.1 | - | 1 | 425 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728151.1 | - | 1 | 204 | Chr. 11 - 80508483 - 80508885 [-] on Build GRCm38 | |

C030013C21Rik
Genomic Map