siRNA Details
| Gene Symbol: Gm9908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079043.1 | - | 1 | 398 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV381911.2 | - | 1 | 406 | Chr. 8 - 44934730 - 44935159 [-] on Build GRCm38 | |

Gm9908
Genomic Map