siRNA Details
| Gene Symbol: 1700063H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006869.1 | - | 2 | 397 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706854.1 | - | 2 | 397 | Chr. 6 - 122391379 - 122392687 [+] on Build GRCm38 | |

1700063H04Rik
Genomic Map