siRNA Details
| Gene Symbol: Mir142hg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA254104.1 | - | 16 | |||
| AI021175.1 | - | 1 | 128 | |||
| AK020764.1 | - | 2 | 52 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mir142hg| Gene Symbol: Mir142hg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA254104.1 | - | 16 | |||
| AI021175.1 | - | 1 | 128 | |||
| AK020764.1 | - | 2 | 52 | |||